Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00238 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024338.2 | 1 | 90 | |||
NR_110307.1 | 1 | 90 | ||||
NR_110308.1 | 1 | 90 | ||||
GenBank mRNA | BC033546.1 | - | 1 | 61 | ||
BC056671.1 | - | 1 | 94 | |||
BC070365.1 | - | 1 | 70 | |||
BI829888.1 | - | 1 | 54 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI520401.1 | - | 1 | 61 | Chr. 14 - 66486400 - 66496849 [+] on Build GRCh38 |