Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFAND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033193.1 | 2, 3 | 118 | |||
NR_033194.1 | 2, 3 | 118 | ||||
NR_033195.1 | 2, 3 | 96 | ||||
NR_033196.1 | 2, 3 | 118 | ||||
GenBank mRNA | AK304111.1 | - | 2, 3 | 101 | ||
BC051857.1 | - | 2, 3 | 100 | 6012307 | ||
BC056877.1 | - | 2, 3 | 101 | 6140929 | ||
BQ435400.1 | - | 2, 3 | 97 |