Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNORA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002952.1 | 1 | 110 | |||
GenBank mRNA | AJ609431.1 | - | 1 | 110 |
Gene Symbol: SNHG15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK096179.1 | - | 1 | 1237 |