Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGT2A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024010.1 | 3 | 981 | |||
GenBank mRNA | AK293381.1 | - | 3 | 552 | ||
BP262099.1 | - | 3 | 279 |