Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAND2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003679.2 | 4 | 722 | |||
NR_136192.1 | 4 | 4128 | ||||
NR_136193.1 | 4 | 4825 | ||||
NR_136194.1 | 4 | 4295 | ||||
NR_136197.1 | 4 | 4199 | ||||
NR_136198.1 | 3 | 4016 | ||||
NR_136199.1 | 5 | 4338 | ||||
NR_136200.1 | 4 | 4828 | ||||
NR_136201.1 | 4 | 4995 | ||||
GenBank mRNA | DQ831955.1 | - | 1 | 2888 | ||
DQ831956.1 | - | 1 | 2888 | |||
EF529495.1 | - | 4 | 3129 | |||
EF529496.1 | - | 4 | 3052 | |||
EF529497.1 | - | 4 | 2999 | |||
EF529498.1 | - | 4 | 3264 | |||
EF529499.1 | - | 4 | 629 | |||
EF529500.1 | - | 4 | 519 | |||
EF529501.1 | - | 3 | 314 | |||
EF529502.1 | - | 3 | 517 | |||
EF529503.1 | - | 4 | 796 | |||
EF529504.1 | - | 5 | 838 | |||
EF529505.1 | - | 4 | 722 | |||
EU000853.1 | - | 4 | 4041 | |||
EU000854.1 | - | 4 | 4041 | |||
EU000855.1 | - | 3 | 5133 | |||
EU000856.1 | - | 3 | 5133 |