Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAND2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003679.2 | 4 | 645 | |||
NR_136192.1 | 4 | 4051 | ||||
NR_136193.1 | 4 | 4748 | ||||
NR_136194.1 | 4 | 4218 | ||||
NR_136197.1 | 4 | 4122 | ||||
NR_136198.1 | 3 | 3939 | ||||
NR_136199.1 | 5 | 4261 | ||||
NR_136200.1 | 4 | 4751 | ||||
NR_136201.1 | 4 | 4918 | ||||
GenBank mRNA | DQ831955.1 | - | 1 | 2811 | ||
DQ831956.1 | - | 1 | 2811 | |||
EF529495.1 | - | 4 | 3052 | |||
EF529496.1 | - | 4 | 2975 | |||
EF529497.1 | - | 4 | 2922 | |||
EF529498.1 | - | 4 | 3187 | |||
EF529499.1 | - | 4 | 552 | |||
EF529500.1 | - | 4 | 442 | |||
EF529501.1 | - | 3 | 237 | |||
EF529502.1 | - | 3 | 440 | |||
EF529503.1 | - | 4 | 719 | |||
EF529504.1 | - | 5 | 761 | |||
EF529505.1 | - | 4 | 645 | |||
EU000853.1 | - | 4 | 3964 | |||
EU000854.1 | - | 4 | 3964 | |||
EU000855.1 | - | 3 | 5056 | |||
EU000856.1 | - | 3 | 5056 |