Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAND2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003679.2 | 1 | 160 | |||
GenBank mRNA | EF529495.1 | - | 1 | 160 | ||
EF529496.1 | - | 1 | 160 | |||
EF529497.1 | - | 1 | 160 | |||
EF529498.1 | - | 1 | 160 | |||
EF529500.1 | - | 1 | 160 | |||
EF529505.1 | - | 1 | 160 | |||
EF529506.1 | - | 1 | 160 | |||
EF529507.1 | - | 1 | 160 | |||
EF529508.1 | - | 1 | 160 | |||
EF529509.1 | - | 1 | 160 |