Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAND2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003679.2 | 4 | 743 | |||
NR_136192.1 | 4 | 4149 | ||||
NR_136193.1 | 4 | 4846 | ||||
NR_136194.1 | 4 | 4316 | ||||
NR_136197.1 | 4 | 4220 | ||||
NR_136198.1 | 3 | 4037 | ||||
NR_136199.1 | 5 | 4359 | ||||
NR_136200.1 | 4 | 4849 | ||||
NR_136201.1 | 4 | 5016 | ||||
GenBank mRNA | DQ831955.1 | - | 1 | 2909 | ||
DQ831956.1 | - | 1 | 2909 | |||
EF529495.1 | - | 4 | 3150 | |||
EF529496.1 | - | 4 | 3073 | |||
EF529497.1 | - | 4 | 3020 | |||
EF529498.1 | - | 4 | 3285 | |||
EF529499.1 | - | 4 | 650 | |||
EF529500.1 | - | 4 | 540 | |||
EF529501.1 | - | 3 | 335 | |||
EF529502.1 | - | 3 | 538 | |||
EF529503.1 | - | 4 | 817 | |||
EF529504.1 | - | 5 | 859 | |||
EF529505.1 | - | 4 | 743 | |||
EU000853.1 | - | 4 | 4062 | |||
EU000854.1 | - | 4 | 4062 | |||
EU000855.1 | - | 3 | 5154 | |||
EU000856.1 | - | 3 | 5154 |