Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAND2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003679.2 | 4 | 624 | |||
NR_136192.1 | 4 | 4030 | ||||
NR_136193.1 | 4 | 4727 | ||||
NR_136194.1 | 4 | 4197 | ||||
NR_136197.1 | 4 | 4101 | ||||
NR_136198.1 | 3 | 3918 | ||||
NR_136199.1 | 5 | 4240 | ||||
NR_136200.1 | 4 | 4730 | ||||
NR_136201.1 | 4 | 4897 | ||||
GenBank mRNA | DQ831955.1 | - | 1 | 2790 | ||
DQ831956.1 | - | 1 | 2790 | |||
EF529495.1 | - | 4 | 3031 | |||
EF529496.1 | - | 4 | 2954 | |||
EF529497.1 | - | 4 | 2901 | |||
EF529498.1 | - | 4 | 3166 | |||
EF529499.1 | - | 4 | 531 | |||
EF529500.1 | - | 3, 4 | 421 | |||
EF529501.1 | - | 2, 3 | 216 | |||
EF529502.1 | - | 3 | 419 | |||
EF529503.1 | - | 4 | 698 | |||
EF529504.1 | - | 5 | 740 | |||
EF529505.1 | - | 4 | 624 | |||
EU000853.1 | - | 4 | 3943 | |||
EU000854.1 | - | 4 | 3943 | |||
EU000855.1 | - | 3 | 5035 | |||
EU000856.1 | - | 3 | 5035 |