Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAND2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003679.2 | 4 | 522 | |||
NR_136192.1 | 4 | 3928 | ||||
NR_136193.1 | 4 | 4625 | ||||
NR_136194.1 | 4 | 4095 | ||||
NR_136197.1 | 4 | 3999 | ||||
NR_136198.1 | 3 | 3816 | ||||
NR_136199.1 | 5 | 4138 | ||||
NR_136200.1 | 4 | 4628 | ||||
NR_136201.1 | 4 | 4795 | ||||
GenBank mRNA | DQ831955.1 | - | 1 | 2688 | ||
DQ831956.1 | - | 1 | 2688 | |||
EF529495.1 | - | 4 | 2929 | |||
EF529496.1 | - | 4 | 2852 | |||
EF529497.1 | - | 4 | 2799 | |||
EF529498.1 | - | 4 | 3064 | |||
EF529499.1 | - | 4 | 429 | |||
EF529502.1 | - | 3 | 317 | |||
EF529503.1 | - | 4 | 596 | |||
EF529504.1 | - | 5 | 638 | |||
EF529505.1 | - | 4 | 522 | |||
EU000853.1 | - | 4 | 3841 | |||
EU000854.1 | - | 4 | 3841 | |||
EU000855.1 | - | 3 | 4933 | |||
EU000856.1 | - | 3 | 4933 |