Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNORA56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002984.1 | 1 | 56 | |||
GenBank mRNA | AJ609449.1 | - | 1 | 56 |
Gene Symbol: DKC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288747.1 | NP_001275676.1 | 12 | 1831 | ||
GenBank mRNA | JF279874.1 | - | 12 | 1654 |