Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNORA81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002989.1 | 1 | 155 | |||
GenBank mRNA | AM055745.1 | - | 1 | 155 |
Gene Symbol: EIF4A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209021.1 | - | 3 | 2202 |