Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC38A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033344.1 | 1 | 292 | |||
XM_017021022.1 | XP_016876511.1 | 1 | 381 | |||
XM_017021025.1 | XP_016876514.1 | 1 | 363 | |||
GenBank mRNA | AF070578.1 | - | 1 | 102 | ||
AK303839.1 | - | 1 | 95 | |||
DC394244.1 | - | 1 | 292 |