Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027139.1 | 5 | 1427 | |||
GenBank mRNA | AK123122.1 | - | 6 | 1621 | ||
BC018073.1 | - | 5 | 1371 | |||
BC032490.1 | - | 5 | 1381 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX375805.2 | - | 3 | 864 | Chr. X - 56976590 - 56984482 [-] on Build GRCh38 |