Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRINS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AK022045.1 | - | LIT3418 PRINS |
1 | 1224 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK024201.1 | - | 1 | 1063 | Chr. 10 - 24247279 - 24251421 [+] on Build GRCh38 |