Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017001115.1 | XP_016856604.1 | 14 | 10653 | ||
XM_017001116.1 | XP_016856605.1 | 15 | 8377 | |||
XM_017001117.1 | XP_016856606.1 | 14 | 10447 | |||
GenBank mRNA | AL138273.1 | - | 1 | 216 | ||
DA733532.1 | - | 1 | 263 |