Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017001115.1 | XP_016856604.1 | 15 | 13359 | ||
XM_017001116.1 | XP_016856605.1 | 16 | 11083 | |||
XM_017001117.1 | XP_016856606.1 | 15 | 13153 | |||
GenBank mRNA | AF448858.1 | - | 1 | 744 | ||
AK054764.1 | - | 1 | 150 | |||
AK095297.1 | - | 2 | 2695 |