Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF448858.1 | - | 1 | 2538 | ||
AK054764.1 | - | 1 | 1944 | |||
BC035431.1 | - | 1 | 734 |