Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRN3P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003369.2 | 17 | 1983 | |||
GenBank mRNA | AK225670.1 | - | 17 | 1833 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM686819.1 | - | 3 | 315 | Chr. 16 - 29104122 - 29116545 [+] on Build GRCh38 |