Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_003245.1 | 2 | 663 | |||
GenBank mRNA | DQ860410.1 | - | LIT3338 HAR1R |
2 | 663 | |
DQ860411.1 | - | LIT3339 HAR1R |
3 | 540 |