Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTTY14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_125733.1 | 5 | 1078 | |||
NR_125734.1 | 4 | 998 | ||||
NR_125735.1 | 3 | 923 | ||||
NR_125736.1 | 5 | 673 | ||||
GenBank mRNA | AF119903.1 | - | LIT2233 CYorf14 |
2 | 1440 |