Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHM1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024386.2 | 3, 4 | 360 | |||
GenBank mRNA | AK301174.1 | - | 3, 4 | 301 | ||
AK304173.1 | - | 4, 5 | 549 | |||
AK308180.1 | - | 4, 5 | 572 | |||
AL833315.1 | - | 3, 4 | 330 | |||
BC034693.1 | - | 4, 5 | 560 |
Gene Symbol: PLEKHM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011525526.1 | XP_011523828.1 | 4, 5 | 603 |