Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHRS4-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023921.2 | 2 | 1724 | |||
NR_023922.2 | 2 | 1465 | ||||
NR_023923.2 | 3 | 1433 | ||||
NR_023924.2 | 2 | 1524 | ||||
GenBank mRNA | BX247982.1 | - | 2 | 1194 | ||
BX647964.1 | - | 3 | 1400 | |||
HQ845197.1 | - | 3 | 1419 | |||
HQ845198.1 | - | 3 | 1419 | |||
HQ845199.1 | - | 2 | 1504 | |||
JQ979294.1 | - | 3 | 1419 | |||
KJ545484.1 | - | 2 | 1359 | |||
KJ574270.1 | - | 2 | 1150 | |||
KJ574271.1 | - | 2 | 1283 | |||
KJ574272.1 | - | 2 | 1283 | |||
KJ574273.1 | - | 2 | 1443 | |||
KJ574274.1 | - | 2 | 1487 | |||
KJ599455.1 | - | 2 | 1217 | |||
KJ599456.1 | - | 2 | 1474 | |||
KJ599457.1 | - | 2 | 1176 | |||
KJ599458.1 | - | 1 | 970 | |||
KJ599459.1 | - | 3 | 1392 | |||
KJ599460.1 | - | 2 | 1518 | |||
KJ599461.1 | - | 3 | 1402 |