Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027783.2 | 4 | 465 | |||
XM_017029720.1 | XP_016885209.1 | 1 | 59 | |||
GenBank mRNA | AK310078.1 | - | 3 | 873 | ||
AK315888.1 | - | 3 | 798 | |||
AL050290.1 | - | 4 | 426 | |||
AY841998.1 | - | 4 | 272 | |||
CD638929.1 | - | 4 | 453 |