Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATXN8OS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_002717.2 | 3 | 748 | |||
GenBank mRNA | AF126749.1 | - | LIT3320 SCA8 |
748 |