Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNQ1DN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024627.1 | 1 | 515 | |||
GenBank mRNA | AB039920.1 | - | 1 | 515 | ||
AJ404617.2 | - | 1 | 485 | |||
BC098159.1 | - | 1 | 377 | |||
BC098256.1 | - | 1 | 377 | |||
BC098319.1 | - | 1 | 381 | |||
BC098344.1 | - | 1 | 377 |