Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SETMAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006713292.3 | XP_006713355.1 | 1 | 220 | ||
XM_006713294.3 | XP_006713357.1 | 1 | 197 | |||
XM_017007032.1 | XP_016862521.1 | 1 | 198 | |||
GenBank mRNA | AK311418.1 | - | 1 | 168 |