Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3GL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301108.1 | NP_001288037.1 | 3, 4 | 494 | ||
NM_001324185.1 | NP_001311114.1 | 4, 5 | 645 | |||
NM_001324187.1 | NP_001311116.1 | 3, 4 | 432 | |||
NR_136711.1 | 4, 5 | 645 | ||||
NR_136713.1 | 3, 4 | 494 | ||||
NR_136714.1 | 3, 4 | 494 | ||||
XM_017022486.1 | XP_016877975.1 | 3, 4 | 599 | |||
GenBank mRNA | AF036270.1 | - | 3, 4 | 191 | ||
AK307442.1 | - | 3, 4 | 346 |