Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP170P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003135.2 | 3, 4 | 274 | |||
GenBank mRNA | BC014590.2 | - | 3, 4 | 274 |
Gene Symbol: CEP170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011544338.2 | XP_011542640.1 | 14, 15 | 5136 | ||
XM_011544342.2 | XP_011542644.1 | 14, 15 | 4978 | |||
XM_017002935.1 | XP_016858424.1 | 15, 16 | 4900 | |||
XM_017002939.1 | XP_016858428.1 | 15, 16 | 4720 | |||
XM_017002940.1 | XP_016858429.1 | 15, 16 | 4363 | |||
XM_017002942.1 | XP_016858431.1 | 15, 16 | 4792 | |||
XM_017002943.1 | XP_016858432.1 | 15, 16 | 4612 | |||
XM_017002945.1 | XP_016858434.1 | 14, 15 | 4656 | |||
XM_017002949.1 | XP_016858438.1 | 14, 15 | 4548 | |||
XM_017002950.1 | XP_016858439.1 | 14, 15 | 4423 |