Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC653653 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027408.1 | 1 | 1097 | |||
GenBank mRNA | AK295685.1 | - | 1 | 1097 |
Gene Symbol: AP1S2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017029925.1 | XP_016885414.1 | 5 | 1119 | ||
GenBank mRNA | AK299921.1 | - | 5 | 966 |