Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001309474.1 | NP_001296403.1 | 1 | 290 | ||
XM_011540210.2 | XP_011538512.1 | 1 | 288 | |||
GenBank mRNA | AK301422.1 | - | 1 | 57 | ||
DC352688.1 | - | 1 | 291 |