Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTX2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029385.1 | 4 | 601 | |||
GenBank mRNA | AB593040.1 | - | 3 | 591 | ||
BC041486.2 | - | 4 | 600 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM799998.1 | - | 4 | 611 | Chr. 14 - 56813183 - 56929380 [+] on Build GRCh38 |