Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTX2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029385.1 | 4 | 831 | |||
GenBank mRNA | AB593040.1 | - | 3 | 821 | ||
BC041486.2 | - | 4 | 830 |