Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033361.1 | 1 | 229 | |||
XM_006713552.3 | XP_006713615.1 | 1 | 274 | |||
GenBank mRNA | AK124671.1 | - | 1 | 230 | ||
BC030619.2 | - | 1 | 147 | |||
BC047517.1 | - | 1 | 147 | |||
DB085398.1 | - | 1 | 154 |