Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNIK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027767.2 | 3 | 570 | |||
GenBank mRNA | BC018093.1 | - | 3 | 567 | ||
BG209385.1 | - | 1 | 106 |