Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQSEC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243197.1 | NP_001230126.1 | 3 | 926 | ||
GenBank mRNA | BC038213.1 | - | 2 | 810 | ||
BC108674.1 | - | 3 | 910 | |||
BX119489.1 | - | 1 | 223 |