Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR100HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024430.2 | 4 | 2833 | |||
NR_137175.1 | 2 | 3104 | ||||
NR_137176.1 | 2 | 2815 | ||||
NR_137177.1 | 2 | 2794 | ||||
NR_137179.1 | 5 | 2787 | ||||
NR_137180.1 | 5 | 2845 | ||||
NR_137181.1 | 3 | 2730 | ||||
NR_137182.1 | 4 | 2812 | ||||
NR_137183.1 | 3 | 2721 | ||||
NR_137184.1 | 2 | 2639 | ||||
NR_137185.1 | 3 | 2700 | ||||
NR_137186.1 | 3 | 2609 | ||||
NR_137187.1 | 3 | 2588 | ||||
NR_137188.1 | 3 | 2678 | ||||
NR_137189.1 | 2 | 3125 | ||||
NR_137190.1 | 3 | 3158 | ||||
NR_137191.1 | 3 | 3137 | ||||
NR_137192.1 | 4 | 2997 | ||||
NR_137193.1 | 5 | 2824 | ||||
NR_137194.1 | 3 | 2699 | ||||
NR_137195.1 | 2 | 2867 | ||||
NR_137196.1 | 2 | 2888 | ||||
GenBank mRNA | AB075498.1 | - | 2 | 3356 | ||
AK021552.1 | - | 1 | 2001 | |||
AK091713.1 | - | 4 | 1637 | |||
AK123947.1 | - | 2 | 3040 | |||
AL832469.1 | - | 3 | 2392 | |||
BC040599.1 | - | 2 | 2418 | |||
BX538058.1 | - | 2 | 2395 | |||
BX647608.1 | - | 2 | 2396 |