Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR100HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024430.2 | 4 | 1133 | |||
NR_137175.1 | 2 | 1404 | ||||
NR_137176.1 | 2 | 1115 | ||||
NR_137177.1 | 2 | 1094 | ||||
NR_137179.1 | 5 | 1087 | ||||
NR_137180.1 | 5 | 1145 | ||||
NR_137181.1 | 3 | 1030 | ||||
NR_137182.1 | 4 | 1112 | ||||
NR_137183.1 | 3 | 1021 | ||||
NR_137184.1 | 2 | 939 | ||||
NR_137185.1 | 3 | 1000 | ||||
NR_137186.1 | 3 | 909 | ||||
NR_137187.1 | 3 | 888 | ||||
NR_137188.1 | 3 | 978 | ||||
NR_137189.1 | 2 | 1425 | ||||
NR_137190.1 | 3 | 1458 | ||||
NR_137191.1 | 3 | 1437 | ||||
NR_137192.1 | 4 | 1297 | ||||
NR_137193.1 | 5 | 1124 | ||||
NR_137194.1 | 3 | 999 | ||||
NR_137195.1 | 2 | 1167 | ||||
NR_137196.1 | 2 | 1188 | ||||
GenBank mRNA | AB075498.1 | - | 2 | 1657 | ||
AK021552.1 | - | 1 | 299 | |||
AK091713.1 | - | 3 | 789 | |||
AK123947.1 | - | 2 | 1341 | |||
AL832469.1 | - | 2 | 721 | |||
BC040599.1 | - | 2 | 717 | |||
BX538058.1 | - | 2 | 695 | |||
BX647608.1 | - | 2 | 695 |