Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THUMPD3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027007.3 | 3 | 1498 | |||
NR_132780.1 | 4 | 1572 | ||||
GenBank mRNA | AL110136.1 | - | 3 | 1494 | ||
BC052961.1 | - | 3 | 3870 |