Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM106CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026810.1 | 1 | 51 | |||
GenBank mRNA | DN989334.1 | - | 1 | 60 |
Gene Symbol: FAM106B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126160.1 | 1 | 51 |