Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291641.1 | NP_001278570.1 | 1, 2 | 123 | ||
XM_017024387.1 | XP_016879876.1 | 1, 2 | 77 | |||
XM_017024392.1 | XP_016879881.1 | 1, 2 | 79 | |||
GenBank mRNA | AL833603.1 | - | 1, 2 | 63 |