Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024393.1 | 1 | 430 | |||
NR_117100.1 | 1 | 430 | ||||
GenBank mRNA | DQ515896.1 | - | 1 | 430 | ||
DQ515897.1 | - | 1 | 430 |