Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024393.1 | 2 | 712 | |||
NR_117100.1 | 2 | 712 | ||||
GenBank mRNA | DQ515896.1 | - | 2 | 712 | ||
DQ515897.1 | - | 2 | 712 | |||
JX003865.1 | - | 1 | 605 |