Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024460.1 | 5, 6 | 780 | |||
XM_017006242.1 | XP_016861731.1 | 5, 6 | 709 | |||
XM_017006243.1 | XP_016861732.1 | 5, 6 | 751 | |||
XM_017006244.1 | XP_016861733.1 | 6, 7 | 790 | |||
GenBank mRNA | AK125303.1 | - | 6, 7 | 877 | ||
AL832692.1 | - | 5, 6 | 720 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB138779.1 | - | 1, 2 | 84 | Chr. 3 - 56566659 - 56593610 [+] on Build GRCh38 |