Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00680-GUSBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003660.2 | 1 | 16 | |||
NR_132997.1 | 1 | 16 | ||||
NR_132998.1 | 1 | 16 | ||||
GenBank mRNA | BM553931.1 | - | 1 | 16 |
Gene Symbol: LINC00680 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125727.1 | 1 | 16 | |||
NR_125728.1 | 1 | 16 | ||||
NR_125729.1 | 1 | 16 | ||||
NR_132994.1 | 1 | 16 | ||||
NR_132995.1 | 1 | 16 | ||||
NR_132996.1 | 1 | 16 |