Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZEB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323638.1 | NP_001310567.1 | 2, 3 | 299 | ||
NM_001323641.1 | NP_001310570.1 | 1, 2 | 343 | |||
NM_001323642.1 | NP_001310571.1 | 1, 2 | 343 | |||
NM_001323646.1 | NP_001310575.1 | 1, 2 | 343 | |||
GenBank mRNA | AK294775.1 | - | 1, 2 | 308 | ||
AK309331.1 | - | 1, 2 | 343 | |||
AK311372.1 | - | 1, 2 | 289 | |||
BX647794.1 | - | 2, 3 | 300 |