Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP32P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003190.2 | 1, 2 | 226 | |||
GenBank mRNA | AK311696.1 | - | 2, 3 | 428 | ||
DA118870.1 | - | 1, 2 | 226 |
Gene Symbol: CCDC144CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK295707.1 | - | 5, 6 | 691 |