Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP32P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003190.2 | 2 | 391 | |||
GenBank mRNA | AK311696.1 | - | 3 | 593 | ||
AK316466.1 | - | 1 | 336 | |||
BC039345.1 | - | 1 | 55 | |||
DA118870.1 | - | 2 | 391 |