Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADORA2A-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028483.2 | 1, 2 | 388 | |||
NR_028484.2 | 1, 2 | 388 | ||||
GenBank mRNA | BC045098.1 | - | 1, 2 | 129 | ||
DA917327.1 | - | 1, 2 | 388 | |||
DB449907.2 | - | 1, 2 | 114 |