Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AQP7P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002817.2 | 4 | 2705 | |||
GenBank mRNA | AK128231.1 | - | 4 | 2701 |
Gene Symbol: AQP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005251453.3 | XP_005251510.1 | 9 | 2809 | ||
XM_006716765.3 | XP_006716828.1 | 7 | 2470 | |||
XM_011517866.2 | XP_011516168.1 | 8 | 2619 | |||
XM_011517867.2 | XP_011516169.1 | 6 | 2249 | |||
XM_017014700.1 | XP_016870189.1 | 8 | 2910 | |||
XM_017014701.1 | XP_016870190.1 | 7 | 2580 | |||
XM_017014702.1 | XP_016870191.1 | 8 | 2615 | |||
XM_017014703.1 | XP_016870192.1 | 7 | 2368 | |||
XM_017014704.1 | XP_016870193.1 | 6 | 2676 | |||
XM_017014705.1 | XP_016870194.1 | 6 | 2346 | |||
XM_017014706.1 | XP_016870195.1 | 8 | 3098 | |||
GenBank mRNA | AK055744.1 | - | 1 | 2655 |